Learn More
Invitrogen™ Phospho-VAV1 (Tyr174) Polyclonal Antibody

Description
Positive Control: Jurkat (serum starvation for 15 min), Jurkat (serum starvation for 15 min, then 0.13 μg/mL anti-T-cell receptor antibody (clone 305) treatment for 5 min) Predicted Reactivity: Rat (100%), Pig (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Phospho-VAV1 (Tyr174) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.23 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol, 1% BSA and 0.025% ProClin 300; pH 7 |
| Gene | VAV1 |
| Gene Accession No. | P15498 |
| Gene Alias | oncogene vav; p95; p95vav; Proto-oncogene vav; VAV; vav 1 guanine nucleotide exchange factor; vav 1 oncogene; vav guanine nucleotide exchange factor 1; vav oncogene; vav proto-oncogene; VAV1; vav-T |
| Gene Symbols | VAV1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.