Learn More
Invitrogen™ Phospho-WAVE1 (Tyr125) Polyclonal Antibody
Description
This antibody was cross-adsorbed to phospho-WAVE (Tyr-150) and unphosphorylated WAVE (Tyr-125) peptides before affinity purification using phospho-WAVE (Tyr-125) peptide. In western blots, the antibody detects an 80 kDa* band corresponding to phosphorylated WAVE in human SYF cSrc transformed cells and mouse macrophages treated with pervanadate, but is not observed in control cells. These bands can be specifically blocked with phospho-WAVE (Tyr-125) peptide (WX1775), but not phospho-WAVE (Tyr-150) peptide (WX1825).
Specifications
Specifications
| Antigen | Phospho-WAVE1 (Tyr125) |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide |
| Gene | WASF1 |
| Gene Accession No. | Q5BJU7, Q8R5H6, Q92558 |
| Gene Alias | AI195380; AI838537; homology of dictyostelium scar 1; KIAA0269; Protein WAVE-1; Scar; SCAR1; Verprolin homology domain-containing protein 1; WAS protein family; WAS protein family member 1; WAS protein family, member 1; WASF1; WASP family 1; WASP family protein member 1; WAVE; WAVE1; WAVE-1; Wiskott-Aldrich syndrome protein family member 1 |
| Gene Symbols | WASF1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.