missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid, TRC

Product Code. 30012066
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012066 50 g 50g
30008415 5 g 5g
30003852 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012066 Supplier Toronto Research Chemicals Supplier No. TRCP35920050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3 O4 P
Molecular Weight (g/mol) 98
Synonym Phosphoric acid, 3M Etching Liquid, Acidum Phosphoricum, Amberphos 54, C 134, C 134 (acid), C 434, C 434 (acid), Conditioner 36, Decon 4512, E 338, EVITs, Etchalite, HQ 54, K Etchant Gel, K-etchant, Kefo, Kerr Etchant, Mikro Klene DF, NSC 80804, Orthophosphoric Acid, PC 400, Panavia Etching Agent, SPA 2, SPA 2 (catalyst), Sonac, TG 434, Total Etch, Ultra-Etch Gel, Ultradent, Ultraetch, Uni-Etch, Vococid, WC-Reiniger, Y 11A06
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Quantity 50 g
InChI Formula InChI=1S/H3O4P/c1-5(2,3)4/h(H3,1,2,3,4)
Formula Weight 97.9769
Percent Purity >95%
Chemical Name or Material Phosphoric Acid (AS)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.