missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phosphoserine/threonine/tyrosine Polyclonal Antibody
GREENER_CHOICE

Product Code. 10352463
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10352463 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10352463 Supplier Invitrogen™ Supplier No. 618300

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody detects pS, pT and pY amino acids with very low cross reactivity to their non-phosphorylated variants.

There are three amino acids that are typically phosphorylated in eukaryotes: serine, threonine, and tyrosine. These phosphoric acid esters can be used as an identifier in the analysis of peptides, proteins, and enzymes. Antibodies that specifically recognize phosphorylated residues have proved to be invaluable to the study of biochemical pathways in which they function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phosphoserine/threonine/tyrosine
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene Alias Phospho-serine; Phospho-threonine; Phospho-tyrosine; pSer; pThr; pTyr; threonine; tyrosine
Host Species Rabbit
Immunogen Phosphothreonine conjugated to protein carriers.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.