Learn More
Phthalate Mixture 392 5-10 μg/mL in Methanol, Dr. Ehrenstorfer
Product Code. 30314262
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30314262 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30314262 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000392ME
This item requires a price update prior to being ordered.
Request a Price for this item here.
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Dibutyl phthalate∣Di-n-octyl phthalate∣Phthalic acid, benzylbutyl ester∣Phthalic acid, bis-2-ethylhexyl ester∣Phthalic acid, bis-cyclohexyl ester∣Phthalic acid, bis-hexyl ester∣Phthalic acid, bis-iso-butyl ester∣Phthalic acid, bis-isodecyl ester∣Phthalic acid, bis-isononyl ester∣Phthalic acid, bis-n-pentyl ester
CAS 84-74-2, 117-84-0, 85-68-7, 117-81-7, 84-61-7, 84-75-3
Quantity 5 mL
Concentration or Composition (by Analyte or Components) Dibutyl phthalate 5 μg/mL∣Di-n-octyl phthalate 5 μg/mL∣Phthalic acid, benzylbutyl ester 5 μg/mL∣Phthalic acid, bis-2-ethylhexyl ester 5 μg/mL∣Phthalic acid, bis-cyclohexyl ester 5 μg/mL∣Phthalic acid, bis-hexyl ester 5 μg/mL∣Phthalic acid, bis-iso-butyl ester 5 μg/mL∣Phthalic acid, bis-n-pentyl ester 5 μg/mL∣Phthalic acid, bis-isodecyl ester 10 μg/mL∣Phthalic acid, bis-isononyl ester 10 μg/mL