missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phthalates Mixture 644 2000 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Dibutyl phthalate∣Diethyl phthalate∣Di-n-octyl phthalate∣Phthalic acid, benzylbutyl ester∣Phthalic acid, bis-2-ethylhexyl ester∣Phthalic acid, bis-cyclohexyl ester∣Phthalic acid, bis-hexyl ester∣Phthalic acid, bis-methyl ester∣Phthalic acid, bis-n-pentyl ester∣Phthalic acid, bis-propyl ester |
| CAS | 84-74-2, 84-66-2, 117-84-0, 85-68-7, 117-81-7, 84-61-7 |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Dibutyl phthalate 2000 μg/mL∣Diethyl phthalate 2000 μg/mL∣Di-n-octyl phthalate 2000 μg/mL∣Phthalic acid, benzylbutyl ester 2000 μg/mL∣Phthalic acid, bis-2-ethylhexyl ester 2000 μg/mL∣Phthalic acid, bis-cyclohexyl ester 2000 μg/mL∣Phthalic acid, bis-hexyl ester 2000 μg/mL∣Phthalic acid, bis-methyl ester 2000 μg/mL∣Phthalic acid, bis-n-pentyl ester 2000 μg/mL∣Phthalic acid, bis-propyl ester 2000 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?