Learn More
Phthalates Mixture 953 1000 μg/mL in Isooctane, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Adipic Acid Bis(2-ethylhexyl) Ester∣Dibutyl phthalate∣Diethyl phthalate∣Di-n-octyl phthalate∣Phthalic acid, benzylbutyl ester∣Phthalic acid, bis-2-ethylhexyl ester∣Phthalic acid, bis-methyl ester |
| CAS | 103-23-1, 84-74-2, 84-66-2, 117-84-0, 85-68-7, 117-81-7 |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Adipic Acid Bis(2-ethylhexyl) Ester 1000 μg/mL∣Dibutyl phthalate 1000 μg/mL∣Diethyl phthalate 1000 μg/mL∣Di-n-octyl phthalate 1000 μg/mL∣Phthalic acid, benzylbutyl ester 1000 μg/mL∣Phthalic acid, bis-2-ethylhexyl ester 1000 μg/mL∣Phthalic acid, bis-methyl ester 1000 μg/mL |