missing translation for 'onlineSavingsMsg'
Learn More

Phthalic acid, bis-2-propylheptyl ester 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30315080
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30315080 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30315080 Supplier Dr. Ehrenstorfer Supplier No. DREA16177040AL100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bis(2-propylheptyl) Phthalate
CAS 53306-54-0
Molecular Formula C28 H46 O4
Quantity 1 mL
Synonym 1,2-Benzenedicarboxylic acid, 1,2-bis(2-propylheptyl) ester, 1,2-Benzenedicarboxylic acid, bis(2-propylheptyl) ester (9CI), Phthalic acid, bis(2-propylheptyl) ester (6CI), Bis(2-propylheptyl) phthalate, DPHP, Di-2-propylheptyl phthalate, Dipropylheptyl phthalate, Hexamoll DPHP, NSC 17071, Palatinol 10P, Phthalic acid, bis-2-propylheptyl ester
InChI Formula InChI=1S/C28H46O4/c1-5-9-11-17-23(15-7-3)21-31-27(29)25-19-13-14-20-26(25)28(30)32-22-24(16-8-4)18-12-10-6-2/h13-14,19-20,23-24H,5-12,15-18,21-22H2,1-4H3
SMILES CCCCCC(CCC)COC(=O)c1ccccc1C(=O)OCC(CCC)CCCCC
IUPAC Name bis(2-propylheptyl) benzene-1,2-dicarboxylate
Molecular Weight (g/mol) 446.66
Formula Weight 446.3396 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.