missing translation for 'onlineSavingsMsg'
Learn More

Phthalic acid, bis-isohexyl ester (mixture of isomers) 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30326479
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30326479 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326479 Supplier Dr. Ehrenstorfer Supplier No. DREA16173580AL100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phthalic acid, bis-iso-hexyl ester
CAS 68515-50-4
Molecular Formula C20H30O4
Quantity 1 mL
Synonym Dihexylphthalate, Phthalic Acid, Bis-isohexyl Ester (Mixture of Isomers), Phthalic acid bis-iso-hexyl ester, 1,2-Benzenedicarboxylic acid, 1,2-dihexyl ester (branched and linear), 1,2-Benzenedicarboxylic acid, dihexyl ester, branched and linear
SMILES CCOC(=O)c1c(cccc1)C(O)=O
Molecular Weight (g/mol) 334.45
Formula Weight 334.21 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.