missing translation for 'onlineSavingsMsg'
Learn More

Phthalic acid, bis-isopentyl ester, Dr. Ehrenstorfer

Product Code. 30314316
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30314316 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30314316 Supplier Dr. Ehrenstorfer Supplier No. DREC16173680
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phthalic acid, bis-iso-pentyl ester
CAS 605-50-5
Molecular Formula C18 H26 O4
Quantity 25 mg
Synonym Phthalic Acid, Bis-isopentyl Ester, 1,2-Benzenedicarboxylic acid, 1,2-bis(3-methylbutyl) ester (ACI), 1,2-Benzenedicarboxylic acid, bis(3-methylbutyl) ester (9CI), Phthalic acid, diisopentyl ester (6CI, 7CI, 8CI), Bis(3-methylbutyl) phthalate, Diisoamyl phthalate, Diisopentyl phthalate, NSC 17070, Palatinol DIPP
InChI Formula InChI=1S/C18H26O4/c1-13(2)9-11-21-17(19)15-7-5-6-8-16(15)18(20)22-12-10-14(3)4/h5-8,13-14H,9-12H2,1-4H3
SMILES CC(C)CCOC(=O)c1ccccc1C(=O)OCCC(C)C
IUPAC Name bis(3-methylbutyl) benzene-1,2-dicarboxylate
Molecular Weight (g/mol) 306.4
Formula Weight 306.1831 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.