missing translation for 'onlineSavingsMsg'
Learn More

Phthalic acid, mixed decyl-hexyl-octyl diester 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30317926
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30317926 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30317926 Supplier Dr. Ehrenstorfer Supplier No. DREA16178600AL100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phthalic acid, mixed decyl-hexyl-octyl diester
CAS 68648-93-1
Quantity 1 mL
Synonym 1,2-Benzenedicarboxylic Acid, Mixed Decyl, Hexyl and Octyl Diesters(1:1:1 Mixture of B185420 and D228455 and D228460)
SMILES O=C(OCCCCCCCC)C1=CC=CC=C1C(OCCCCCC)=O.O=C(OCCCCCCCCCC)C2=CC=CC=C2C(OCCCCCC)=O.O=C(OCCCCCCCCCC)C3=CC=CC=C3C(OCCCCCCCC)=O
Molecular Weight (g/mol) 390.556
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.