missing translation for 'onlineSavingsMsg'
Learn More

Phthalic acid, mono-2-ethylhexyl ester 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30318193
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30318193 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30318193 Supplier Dr. Ehrenstorfer Supplier No. DREA16178900AL100
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phthalic Acid Mono-2-ethylhexyl Ester
CAS 4376-20-9
Molecular Formula C16 H22 O4
Quantity 1 mL
Synonym Phthalic acid, mono-2-ethylhexyl ester
InChI Formula InChI=1S/C16H22O4/c1-3-5-8-12(4-2)11-20-16(19)14-10-7-6-9-13(14)15(17)18/h6-7,9-10,12H,3-5,8,11H2,1-2H3,(H,17,18)
SMILES CCCCC(CC)COC(=O)c1ccccc1C(=O)O
IUPAC Name 2-(2-ethylhexoxycarbonyl)benzoic acid
Molecular Weight (g/mol) 278.34
Formula Weight 278.1518 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.