missing translation for 'onlineSavingsMsg'
Learn More

Phthalic acid, n-dodecyl-n-undecyl ester, Dr. Ehrenstorfer

Product Code. 30317846
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30317846 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30317846 Supplier Dr. Ehrenstorfer Supplier No. DREC16178200
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phthalic Acid, n-Dodecyl-n-Undecyl ester
CAS 116998-09-5
Molecular Formula C31 H52 O4
Quantity 50 mg
Synonym 1,2-Benzenedicarboxylic acid, dodecyl undecyl ester (9CI), Dodecyl undecyl phthalate, Jayflex UDP
InChI Formula InChI=1S/C31H52O4/c1-3-5-7-9-11-13-15-17-19-23-27-35-31(33)29-25-21-20-24-28(29)30(32)34-26-22-18-16-14-12-10-8-6-4-2/h20-21,24-25H,3-19,22-23,26-27H2,1-2H3
SMILES CCCCCCCCCCCCOC(=O)c1ccccc1C(=O)OCCCCCCCCCCC
IUPAC Name 2-O-dodecyl 1-O-undecyl benzene-1,2-dicarboxylate
Molecular Weight (g/mol) 488.7422
Formula Weight 488.3866 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.