missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PIGN Polyclonal Antibody
GREENER_CHOICE

Product Code. 15706861 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15706861 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15706861 Supplier Invitrogen™ Supplier No. PA558987

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: SWGISHTRVP TESRPGHVAL IAGFYEDVSA VAKGWKENPV EFDSLFNESK YTWSWGSPDI LPMFAKGASG DHVYTYSYDA KREDFGAQDA TKLDTW Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

PIGN is involved in glycosylphosphatidylinositol (GPI)-anchor biosynthesis. The GPI-anchor is a glycolipid found on many blood cells and serves to anchor proteins to the cell surface. This protein is expressed in the endoplasmic reticulum and transfers phosphoethanolamine (EtNP) to the first mannose of the GPI anchor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIGN
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PIGN
Gene Accession No. O95427
Gene Alias Gm20308; GPI ethanolamine phosphate transferase 1; MCAHS; MCAHS1; MCD4; MCD4 homolog; MDC4; phosphatidylinositol glycan anchor biosynthesis class N; phosphatidylinositol glycan anchor biosynthesis, class N; phosphatidylinositol glycan, class N; phosphatidylinositol-glycan biosynthesis class N protein; PIGN; PIG-N
Gene Symbols PIGN
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PIGN. Recombinant protein control fragment (Product #RP-98291).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23556
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.