missing translation for 'onlineSavingsMsg'
Learn More

PIKFyve/PIP5K3 Antibody, Novus Biologicals™

Product Code. 18699256 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18699256 100 μg 100µL
18680287 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18699256 Supplier Novus Biologicals Supplier No. NBP312608100ul

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PIKFyve/PIP5K3 Polyclonal antibody specifically detects PIKFyve/PIP5K3 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIKFyve/PIP5K3
Applications Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunoprecipitation 1-5uL/mg of uLysate, Immunohistochemistry-Paraffin 1:200-1:1000
Formulation PBS, pH7.0
Gene Alias 1-phosphatidylinositol-3-phosphate 5-kinase, EC 2.7.1.150, FLJ37746, FYVE finger-containing phosphoinositide kinase, KIAA0981CFD, MGC40423, p235, Phosphatidylinositol-3-phosphate 5-kinase, Phosphatidylinositol-3-phosphate 5-kinase type III, phosphoinositide kinase, FYVE finger containing, PIKfyve, PIP5K, PIP5K3FAB1, PIPkin-III, type III, Type III PIP kinase
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PIKFyve/PIP5K3 fragment
Purification Method Antigen and protein A Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 200576
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.