missing translation for 'onlineSavingsMsg'
Learn More

Piperonylonitrile, 97%, Thermo Scientific™

Product Code. 10735991 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10735991 25 g Glass bottle 25g
10103110 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10735991 Supplier Thermo Scientific Acros Supplier No. 151880250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 4421-09-4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00005820
InChI Key PKRWWZCDLJSJIF-UHFFFAOYSA-N
Synonym piperonylonitrile, benzo d 1,3 dioxole-5-carbonitrile, 3,4-methylenedioxy benzonitrile, 3,4-methylenedioxybenzonitrile, 2h-1,3-benzodioxole-5-carbonitrile, benzo-1,3-dioxole-5-carbonitrile, benzonitrile, 3,4-methylenedioxy, 2h-benzo d 1,3-dioxolane-5-carbonitrile, pubchem15358
PubChem CID 78136
IUPAC Name 1,3-benzodioxole-5-carbonitrile
SMILES C1OC2=C(O1)C=C(C=C2)C#N

Specifications

Melting Point 90°C to 95°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C8H5NO2
Quantity 25 g
Beilstein 19, 270
Formula Weight 147.13
Percent Purity ≥96.0%
Physical Form Crystalline Powder or Needles
Chemical Name or Material Piperonylonitrile, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.