missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PLD6 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15537152
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15537152 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15537152 Supplier Invitrogen™ Supplier No. PA546348

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: ITEDDEYVRL FLEEFERIWE QFNPTKYTFF PPKKSHGSCA PPVSRAGGRL Sequence homology: Human: 100%.

PLD6 is a cardiolipin hydrolase present at the mitochondrial outer membrane required both for mitochondrial fusion and piRNA metabolic process. PLD6 acts by catalyzing the hydrolysis of cardiolipin (diphosphatidylglycerol) to form phosphatidate (phosphatidic acid or PA) at the mitochondrial outer membrane surface, promoting mitochondrial fusion. The production of phosphatidate also regulates the piRNA metabolic process by promoting recruitment and/or activation of components of the meiotic nuage, also named P granule, a critical step for primary biogenesis of piRNAs. It is required during spermatogenesis to repress transposable elements and prevent their mobilization via its role in the piRNA metabolic process, which mediates the repression of transposable elements during meiosis by forming complexes composed of piRNAs and Piwi proteins and govern the methylation and subsequent repression of transposons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLD6
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PLD6
Gene Accession No. Q8N2A8
Gene Alias 4933433K01Rik; choline phosphatase 6; Gm10; Mitochondrial cardiolipin hydrolase; mitochondrial phospholipase; mitoPLD; mZuc; novel Phospholipase D Active site motif-containing protein; phosphatidylcholine-hydrolyzing phospholipase D6; phospholipase D family member 6; phospholipase D family, member 6; phospholipase D6; PLD 6; PLD6; Protein zucchini homolog; Zuc
Gene Symbols PLD6
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human PLD6 (aa 188-237).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201164
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.