missing translation for 'onlineSavingsMsg'
Learn More

PLEKHB1/PHR1 Antibody [HRP], Novus Biologicals™

Product Code. 30749384 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30749384 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30749384 Supplier Novus Biologicals Supplier No. NBP347119H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PLEKHB1/PHR1 Polyclonal antibody specifically detects PLEKHB1/PHR1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLEKHB1/PHR1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias Evectin-1, EVT1, KPL1evectin-1, PH domain containing, retinal 1, PH domain-containing family B member 1, PH domain-containing protein in retina 1, PHR1PH domain containing protein in retina 1, PHRET1, pleckstrin homology domain containing, family B (evectins) member 1, Pleckstrin homology domain retinal protein 1, pleckstrin homology domain-containing family B member 1
Host Species Rabbit
Immunogen pleckstrin homology domain containing, family B(evectins) member 1. (Uniprot# Q9UF11)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58473
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.