missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Plexin-A1 Recombinant Rabbit Monoclonal Antibody (24GB1245)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557609
This item is not returnable.
View return policy
Description
Plexin-A1 Recombinant Monoclonal Antibody for Western Blot
Coreceptor for SEMA3A, SEMA3C, SEMA3F and SEMA6D. Necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. Plays a role in axon guidance, invasive growth and cell migration. Class 3 semaphorins bind to a complex composed of a neuropilin and a plexin. The plexin modulates the affinity of the complex for specific semaphorins, and its cytoplasmic domain is required for the activation of down-stream signaling events in the cytoplasm. [UniProt].
Specifications
| Plexin-A1 | |
| Recombinant Monoclonal | |
| 0.7 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P70206, Q9UIW2 | |
| PLXNA1 | |
| A synthesized peptide derived from human Plexin A1 (aa 100-180). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot | |
| 24GB1245 | |
| Unconjugated | |
| PLXNA1 | |
| 2600013D04Rik; Kiaa4053; mKIAA4053; NOV; NOVP; Plex 1; PlexA1; plexin 1; plexin A1; plexin-1; plexin-A1; PLXN1; Plxna1; RGD1560871; semaphorin receptor NOV | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18844, 362398, 5361 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction