missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Plexin-A1 Recombinant Rabbit Monoclonal Antibody (24GB1245)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

198.00€ - 564.00€

Specifications

Antigen Plexin-A1
Clone 24GB1245
Concentration 0.7 mg/mL
Content And Storage -20°C
Applications Flow Cytometry, Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30275302
GREENER_CHOICE
View Documents
Invitrogen™
MA557608
100 μL
564.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30275534
View Documents
Invitrogen™
MA557609
20 μL
198.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Plexin-A1 Recombinant Monoclonal Antibody for Western Blot

Coreceptor for SEMA3A, SEMA3C, SEMA3F and SEMA6D. Necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. Plays a role in axon guidance, invasive growth and cell migration. Class 3 semaphorins bind to a complex composed of a neuropilin and a plexin. The plexin modulates the affinity of the complex for specific semaphorins, and its cytoplasmic domain is required for the activation of down-stream signaling events in the cytoplasm. [UniProt]
Specifications

Specifications

Plexin-A1
0.7 mg/mL
Flow Cytometry, Western Blot
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P70206, Q9UIW2
18844, 362398, 5361
A synthesized peptide derived from human Plexin A1 (aa 100-180).
Antibody
24GB1245
-20°C
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
PLXNA1
2600013D04Rik; Kiaa4053; mKIAA4053; NOV; NOVP; Plex 1; PlexA1; plexin 1; plexin A1; plexin-1; plexin-A1; PLXN1; Plxna1; RGD1560871; semaphorin receptor NOV
PLXNA1
Primary
Affinity chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.