missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PMAT Polyclonal Antibody
GREENER_CHOICE

Product Code. 15654805
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15654805 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15654805 Supplier Invitrogen™ Supplier No. PA550877

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total PMAT protein.

PMAT(Slc29a4) is a member of the SLC29 family and encodes a plasma membrane protein with 11 transmembrane helices. This protein catalyzes the re-uptake of monoamines into presynaptic neurons, thus determining the intensity and duration of monoamine neural signaling. It has been shown to transport several compounds, including serotonin, dopamine, and the neurotoxin 1-methyl-4-phenylpyridinium.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PMAT
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene SLC29A4
Gene Accession No. Q7RTT9
Gene Alias ENT4; Equilibrative nucleoside transporter 4; FLJ34923; hENT4; mPMAT; Plasma membrane monoamine transporter; PMAT; PSEC0113; SLC29A4; solute carrier family 29 (equilibrative nucleoside transporter), member 4; solute carrier family 29 (nucleoside transporters), member 4; solute carrier family 29 member 4
Gene Symbols SLC29A4
Host Species Rabbit
Immunogen Fusion protein corresponding to a region derived from internal residues of human solute carrier family 29 (equilibrative nucleoside transporter), member 4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222962
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.