missing translation for 'onlineSavingsMsg'
Learn More

PMS2 Mouse anti-Human, Mouse, Rat, Clone: 1G4E6, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66075-1-IG-150UL

 View more versions of this product

Product Code. 16807625

  • 410.00€ / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PMS1 protein homologue 2 (PMS2) is a DNA mismatch repair protein. Defects in PMS2 are a cause of hereditary non-polyposis cancer (HNPCC). Human PMS2 is an amino acid protein of 862 amino acids.
Specifications

Specifications

PMS2
Monoclonal
1.58 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P54278, P54279
Pms2
PMS2 Fusion Protein Ag12661
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG2a
Immunohistochemistry (Paraffin), Western Blot
1G4E6
Unconjugated
PMS2
HNPCC4, PMS1 protein homolog 2, PMS2, PMS2CL, PMSL2
Mouse
Protein A
RUO
18861, 288479, 5395
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.