missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PMS2 Mouse anti-Human, Mouse, Rat, Clone: 1G4E6, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | PMS2 |
|---|---|
| Clone | 1G4E6 |
| Concentration | 1.58 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16817625
|
Proteintech
66075-1-IG-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16807625
|
Proteintech
66075-1-IG-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PMS1 protein homologue 2 (PMS2) is a DNA mismatch repair protein. Defects in PMS2 are a cause of hereditary non-polyposis cancer (HNPCC). Human PMS2 is an amino acid protein of 862 amino acids.Specifications
| PMS2 | |
| 1.58 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| HNPCC4, PMS1 protein homolog 2, PMS2, PMS2CL, PMSL2 | |
| Pms2 | |
| IgG2a | |
| Protein A | |
| Antibody |
| 1G4E6 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Mouse, Rat, Human | |
| P54278, P54279 | |
| 18861, 288479, 5395 | |
| PMS2 Fusion Protein Ag12661 | |
| Primary | |
| -20°C | |
| PMS2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title