missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNPT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14487-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody detected the endogenous PNPT1 around 80 kDa in mouse brain.
PNPase is involved with the exosome complex, which is involved in 3-prime-to-5-prime exoribonuclease activity for RNA processing and degradation.PNPT1 is a subunit of the exosome complex, which is involved in 3-prime-to-5-prime exoribonuclease activity for RNA processing and degradation.Specifications
| PNPT1 | |
| Polyclonal | |
| Unconjugated | |
| PNPT1 | |
| 3 5 RNA exonuclease OLD35, old 35, OLD35, PNPASE, PNPase 1, PNPase old 35, PNPT1, Polynucleotide phosphorylase 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 360992, 71701, 87178 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8K1R3, Q8TCS8 | |
| Pnpt1 | |
| PNPT1 Fusion Protein Ag5888 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction