missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNPT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | PNPT1 |
|---|---|
| Concentration | 0.37 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16873144
|
Proteintech
14487-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16863144
|
Proteintech
14487-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody detected the endogenous PNPT1 around 80 kDa in mouse brain.
PNPase is involved with the exosome complex, which is involved in 3-prime-to-5-prime exoribonuclease activity for RNA processing and degradation.PNPT1 is a subunit of the exosome complex, which is involved in 3-prime-to-5-prime exoribonuclease activity for RNA processing and degradation.Specifications
| PNPT1 | |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human, Rat, Mouse | |
| Q8K1R3, Q8TCS8 | |
| 360992, 71701, 87178 | |
| PNPT1 Fusion Protein Ag5888 | |
| Primary | |
| -20°C | |
| PNPT1 |
| 0.37 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| 3 5 RNA exonuclease OLD35, old 35, OLD35, PNPASE, PNPase 1, PNPase old 35, PNPT1, Polynucleotide phosphorylase 1 | |
| Pnpt1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title