missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10288-1-AP-150UL
This item is not returnable.
View return policy
Description
The function of the small subunit is not yet clear.Specifications
| POLD2 | |
| Polyclonal | |
| Unconjugated | |
| POLD2 | |
| DNA polymerase delta subunit 2, POLD2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18972, 289758, 5425 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O35654, P49005, Q6AXY4 | |
| Pold2 | |
| POLD2 Fusion Protein Ag0270 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction