Learn More
POLD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10288-1-AP-20UL
 View more versions of this product
Product Code. 16896573
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
The function of the small subunit is not yet clear.
Specifications
Specifications
POLD2
Polyclonal
Unconjugated
POLD2
DNA polymerase delta subunit 2, POLD2
Rabbit
Antigen Affinity Chromatography
RUO
18972, 289758, 5425
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O35654, P49005, Q6AXY4
Pold2
POLD2 Fusion Protein Ag0270
20 μL
Primary
Rat, Human, Mouse