missing translation for 'onlineSavingsMsg'
Learn More

POLN Antibody (1G2/2), Biotin, Novus Biologicals™

Product Code. 18668078 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18668078 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18668078 Supplier Novus Biologicals Supplier No. NBP250253B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

POLN Monoclonal antibody specifically detects POLN in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLN
Applications Western Blot
Classification Monoclonal
Clone 1G2/2
Conjugate Biotin
Formulation PBS
Gene Alias DNA polymerase N, DNA polymerase nu, DNA polymerase POL4P, EC 2.7.7.7, POL4P, polymerase (DNA directed) nu
Host Species Mouse
Immunogen Recombinant fragment of mouse POLN protein
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 353497
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.