missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TAG-72 Monoclonal antibody specifically detects TAG-72 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | TAG-72 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | B72.3 (Satumomab) |
| Conjugate | APC-Cyanine7 |
| Formulation | PBS |
| Gene Alias | CA 72-4, Protein TAG-72, tag 72, tag-72 Protein TAG-72 |
| Host Species | Rabbit |
| Immunogen | Tumour-associated glycoprotein 72 is a glycoprotein found on the surface of many cancer cells, including ovary, breast, colon, and pancreatic cells. |
| Purification Method | Protein A purified |
| Show More |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?