missing translation for 'onlineSavingsMsg'
Learn More

Potassium Ferrocyanide Trihydrate ACS MP Biomedicals

Product Code. 11457842 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11457842 100 g 100g
11467842 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11457842 Supplier MP Biomedicals Supplier No. 0215256080
  • ONLINE EXCLUSIVE PRICE
    45.20€
    43.05€ / 100g
    Save 2.15€
    4% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 14459-95-1
Molecular Formula C6H6FeK4N6O3
Molecular Weight (g/mol) 422.39
MDL Number MFCD00167023
InChI Key NLYKFUFIUWPPIZ-UHFFFAOYSA-N
Synonym Potassium Hexacyanoferate II
IUPAC Name tetrapotassium hexacyanoironbis(ylium) trihydrate
SMILES O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Quantity 100 g
Formula Weight 422.39
Grade ACS
Density 7.87 g/cm3 at 25 °C
Chemical Name or Material Potassium Ferrocyanide, Trihydrate
compliance-icons
Product Identifier
  • Potassium Ferrocyanide ACS Grade
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.