Learn More
Potassium hexacyanoferrate(II) trihydrate, 98+%
Description
Potassium hexacyanoferrate(II) trihydrate is widely used in anticaking agent for both road and table salt. It is involved in the purification of tin and separation of copper from molybdenum ores. It plays a vital role in the in production of wine and citric acid. In analytical chemistry, it is used to determine the concentration of potassium permanganate. It is in combination with potassium ferricyanide and phosphate buffered solution acts as buffer for beta-galatosidase. Further, it is used as fertilizers for plants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Potassium hexacyanoferrate(II) trihydrate is widely used in anticaking agent for both road and table salt. It is involved in the purification of tin and separation of copper from molybdenum ores. It plays a vital role in the in production of wine and citric acid. In analytical chemistry, it is used to determine the concentration of potassium permanganate. It is in combination with potassium ferricyanide and phosphate buffered solution acts as buffer for beta-galatosidase. Further, it is used as fertilizers for plants.
Solubility
Soluble in water. Insoluble in ethanol and ether.
Notes
Begins to lose water at 600C, anhydrous at 1000C. Incompatible with oxidizing agents and strong acids.
Specifications
Specifications
| Melting Point | 69°C to 71°C |
| pH | 9.5 |
| Quantity | 2500 g |
| Merck Index | 14,7631 |
| Solubility Information | Soluble in water. Insoluble in ethanol and ether. |
| Formula Weight | 422.39 (368.36 Anhydrous) |
| Percent Purity | ≥98% |
| Odor | Odorless |
| Density | 1.85 |
| Chemical Name or Material | Potassium hexacyanoferrate(II) trihydrate |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.