Learn More
Potassium Hydroxide, 0.200 N (N/5) in Isopropyl Alcohol, Ricca Chemical
Product Code. 31940493
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
Unit Size:
1 L
4 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
31940493 500 mL Natural Poly Bottle 500mL
31940466 1 L Natural Poly Bottle 1 L
31940512 4 L Natural Poly Bottle 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 31940493 Supplier Ricca Chemical Company Supplier No. R6254000500A
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
Chemical Identifiers
CAS 67-63-0 , 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]
Specifications
Color Colorless
Assay Percent Range 0.2000 ± 0.0005 N at 20°C
Solubility Information Solubility in Water: Infinite
Quantity 500 mL
Grade Laboratory
Odor Characteristic alcohol odor
Packaging Natural Poly Bottle
Solvent or Matrix Nonaqueous Solution
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide, 0.2N in Isopropyl Alcohol