Learn More
Potassium Hydroxide, 0.200 N (N/5) in Isopropyl Alcohol, Ricca Chemical
Product Code. 31940512
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
Unit Size:
1 L
4 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
31940512 4 L Natural Poly Bottle 4 L
31940493 500 mL Natural Poly Bottle 500mL
31940466 1 L Natural Poly Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 31940512 Supplier Ricca Chemical Company Supplier No. R62540004A
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket
Chemical Identifiers
CAS 67-63-0 , 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]
Specifications
Color Colorless
Assay Percent Range 0.2000 ± 0.0005 N at 20°C
Solubility Information Solubility in Water: Infinite
Quantity 4 L
Grade Laboratory
Odor Characteristic alcohol odor
Packaging Natural Poly Bottle
Solvent or Matrix Nonaqueous Solution
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide, 0.2N in Isopropyl Alcohol