missing translation for 'onlineSavingsMsg'
Learn More

Pralidoxime Chloride, TRC

Product Code. 30425270
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30425270 1 g 1g
30428236 10 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30425270 Supplier Toronto Research Chemicals Supplier No. TRCP7011201G
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-15-0
Molecular Formula C7 H9 N2 O . Cl
Molecular Weight (g/mol) 172.61
Synonym Pyridinium, 2-[(hydroxyimino)methyl]-1-methyl-, chloride (1:1), 2-Formyl-1-methylpyridinium chloride oxime (6CI), Pyridinium, 2-[(hydroxyimino)methyl]-1-methyl-, chloride (9CI), Pyridinium, 2-formyl-1-methyl-, chloride, oxime (8CI), 1-Methyl-2-aldoximinopyridinium chloride, 1-Methylpyridinium-2-aldoxime chloride, 2-(Hydroxyiminomethyl)-1-methylpyridinium chloride, 2-PAM chloride, 2-Pralidoxime chloride, 2-Pyridinealdoxime methochloride, N-Methylpyridinium-2-aldoxime chloride, Pralidoxime chloride, Protopam chloride
IUPAC Name (NE)-N-[(1-methylpyridin-1-ium-2-yl)methylidene]hydroxylamine;chloride
SMILES [Cl-].C[n+]1ccccc1\C=N\O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H8N2O.ClH/c1-9-5-3-2-4-7(9)6-8-10;/h2-6H,1H3;1H
Formula Weight 172.0403
Chemical Name or Material Pralidoxime chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.