missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PRELP Polyclonal specifically detects PRELP in Human samples. It is validated for ELISA.
Specifications
Specifications
| Classification | Polyclonal |
| Formulation | PBS |
| Host Species | Rabbit |
| Quantity | 0.1 mL |
| Primary or Secondary | Primary |
| Target Species | Human |
| Content And Storage | Store at 4C in the dark. |
| Product Type | Antibody |
| Form | Purified |
| Product Line | Novus |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Vous avez repéré une opportunité d'amélioration ?