missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PREP Polyclonal Antibody
GREENER_CHOICE

Product Code. 17208583 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17208583 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17208583 Supplier Invitrogen™ Supplier No. PA5115798

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Prolyl Endopeptidase.

p33cdk2 associates with cyclin A in human cells. Kinase activity associated with cyclin A-cdc2 is found only in G2-phase. Cdk2 also complexes with cyclins E, D1, and D3. Cyclin E-cdk2 kinase is active in the G1- and S-phases of the cell cycle and is important (as does cyclin A-cdk2) for the progression from G1-to S-phase. The levels of cyclin A-cdk2 is maximal at the G1/S transition and both cdk2 and cyclin A are found associated with DNA in the initiation complex during replication. Rb protein acts as substrate for cdk2-cyclin E and/or cdk2-cyclin A in vivo. Cdk2 is activated and specifically localized to the nucleus during late G1-, S-, and G2-phase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PREP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene PREP
Gene Accession No. O70196, P48147, Q9QUR6
Gene Alias AI047692; AI450383; D10Wsu136e; dJ355L5.1 (prolyl endopeptidase); EC 3.4.21.26; MGC16060; PE; PE {ECO:0000250; PEP; Pop; post-proline cleaving enzyme; post-proline cleaving enzyme {ECO:0000250; Prep; prep {ECO:0000312; Prolyl endopeptidase; prolyl endopeptidase {ECO:0000250; prolyl oligopeptidase; RGD:620841}; rPop; UniProtKB:P23687}
Gene Symbols PREP
Host Species Rabbit
Immunogen A synthesized peptide derived from human PREP(Accession P48147), corresponding to amino acid residues L168-D218.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19072, 5550, 83471
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.