Learn More
PRKACA Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24503-1-AP-150UL
 View more versions of this product
Product Code. 16872975
  • 410.00€ / 150µL
Description
The PRKACA antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human PRKACA. This antibody recognizes human, mouse, rat antigen. The PRKACA antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
PRKACA
Polyclonal
Unconjugated
150 μL
5566
IgG
Western Blot, Immunohistochemistry, ELISA
333 μg/mL
Rabbit
Primary
Human, Rat, Mouse