missing translation for 'onlineSavingsMsg'
Learn More

PRKACA Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Specifications

Antigen PRKACA
Concentration 333 μg/mL
Applications Western Blot, Immunohistochemistry, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16882975
View Documents
Proteintech
24503-1-AP-20UL
20 μL
145.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16872975
View Documents
Proteintech
24503-1-AP-150UL
150 μL
410.00€
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The PRKACA antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human PRKACA. This antibody recognizes human, mouse, rat antigen. The PRKACA antibody has been validated for the following applications: IHC, WB, ELISA analysis.

Specifications

Specifications

PRKACA
Western Blot, Immunohistochemistry, ELISA
Unconjugated
Human, Rat, Mouse
IgG
333 μg/mL
Polyclonal
Rabbit
5566
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.