missing translation for 'onlineSavingsMsg'
Learn More

PRKX Antibody [Biotin], Novus Biologicals™

Product Code. 30758969 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30758969 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30758969 Supplier Novus Biologicals Supplier No. NBP346891B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PRKX Polyclonal antibody specifically detects PRKX in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRKX
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias EC 2.7.11, PKX1EC 2.7.11.1, Protein kinase PKX1, protein kinase, X-linked, serine/threonine-protein kinase PRKX
Host Species Rabbit
Immunogen protein kinase, X-linked. (Uniprot# P51817)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5613
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.