Learn More
PRMT5 Antibody (RAB-C136) - F(ab) - Azide and BSA Free, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals NBP3-20130-0.025mg
 View more versions of this product
Product Code. 18698034
  • 399.00€ / 0.02mg
Description
PRMT5 Monoclonal antibody specifically detects PRMT5 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP assay (ChIP)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PRMT5
Monoclonal
1 mg/mL
PBS
Human
Affinity purified
RUO
Primary
Human
Purified
ELISA, Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP Assay
RAB-C136
Unconjugated
EC 2.1.1.-, EC 2.1.1.125, Histone-arginine N-methyltransferase PRMT5, HMT1 hnRNP methyltransferase-like 5,72 kDa ICln-binding protein, HRMT1L5, IBP72, Jak-binding protein 1, JBP1, protein arginine methyltransferase 5, protein arginine N-methyltransferase 5, Shk1 kinase-binding protein 1 homolog, SKB1, skb1 (S. pombe) homolog, SKB1 homolog (S. pombe), SKB1HsSKB1 homolog
This antibody was obtained by recombinant antibody (rAb) phage display recognizing PRMT5 protein under non-denaturing conditions.
0.025 mg
Cell Cycle and Replication, Chromatin Research, Circadian Rhythm
10419
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.