missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRMT5 Antibody (RAB-C136) - F(ab) - Azide and BSA Free, Novus Biologicals™
Human Monoclonal Antibody
399.00€ - 849.00€
Specifications
| Antigen | PRMT5 |
|---|---|
| Clone | RAB-C136 |
| Concentration | 1 mg/mL |
| Applications | ELISA, Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP Assay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18698034
|
Novus Biologicals
NBP3-20130-0.025mg |
0.025 mg |
399.00€
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18618454
|
Novus Biologicals
NBP3-20130-0.1mg |
0.1 mg |
849.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PRMT5 Monoclonal antibody specifically detects PRMT5 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP assay (ChIP)Specifications
| PRMT5 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.1.1.-, EC 2.1.1.125, Histone-arginine N-methyltransferase PRMT5, HMT1 hnRNP methyltransferase-like 5,72 kDa ICln-binding protein, HRMT1L5, IBP72, Jak-binding protein 1, JBP1, protein arginine methyltransferase 5, protein arginine N-methyltransferase 5, Shk1 kinase-binding protein 1 homolog, SKB1, skb1 (S. pombe) homolog, SKB1 homolog (S. pombe), SKB1HsSKB1 homolog | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing PRMT5 protein under non-denaturing conditions. | |
| Affinity purified |
| RAB-C136 | |
| ELISA, Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP Assay | |
| Unconjugated | |
| Human | |
| Cell Cycle and Replication, Chromatin Research, Circadian Rhythm | |
| PBS | |
| 10419 | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title