missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Properdin Antibody (Novelmed patent anti-Properdin) [Janelia Fluor™ 635], Novus Biologicals™
Description
Properdin Monoclonal antibody specifically detects Properdin in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
Specifications
| Antigen | Properdin |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Clone | Novelmed patent anti-Properdin |
| Conjugate | Janelia Fluor 635 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | BFD, Complement factor P, complement factor properdin, PFCcomplement, PFD, PROPERDIN |
| Host Species | Human |
| Immunogen | Complement Factor P/ Properdin |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?