Learn More
Propionic anhydride, 99
Product Code. 10563681 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Unit Size:
1 L
2.5 L
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10563681 1 L 1 L
10669783 250 mL 250mL
10300844 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10563681 Supplier Thermo Scientific Acros Supplier No. 131520010
 Restricted Item
In Stock
Add to basket
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
CAS: 123-62-6 | C6;H1;0;O3; | 130.14 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 123-62-6
Molecular Formula C6;H1;0;O3;
Molecular Weight (g/mol) 130.14
Specifications
Melting Point -42.0°C
Density 1.0150 g/mL
Boiling Point 167.0°C to 170.0°C
Flash Point 63°C
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4030 to 1.4050
Quantity 1 L
Beilstein 02, 242
Merck Index 15, 7940
Specific Gravity 1.0150
Solubility Information Solubility aq. soln.: hydrolyses aq. soln. forming propionic acid. Other solubilities: soluble in methanol, ethanol, ether and chloroform
Viscosity 1.144 mPa.s (20°C)
Formula Weight 130.14
Chemical Name or Material Propionic anhydride
Show More Show Less
compliance-icons
Product Identifier
  • Propionic anhydride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Propionic anhydride