missing translation for 'onlineSavingsMsg'
Learn More

Propyl p-toluenesulfonate, 98+%

Product Code. 10066022 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10066022 25 g Glass bottle 25g
10491523 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10066022 Supplier Thermo Scientific Acros Supplier No. 380600250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 599-91-7 | C10H14O3S | 214.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 599-91-7
Molecular Formula C10H14O3S
Molecular Weight (g/mol) 214.28
MDL Number MFCD00059244
InChI Key JTTWNTXHFYNETH-UHFFFAOYSA-N
Synonym propyl p-toluenesulfonate, propyl tosylate, benzenesulfonic acid, 4-methyl-, propyl ester, p-toluenesulfonic acid, propyl ester, propyl toluene-4-sulphonate, ccris 9150, p-toluenesulfonic acid n-propyl ester, p-toluenesulfonic acid propyl ester, propanol tosylate, acmc-209mha
PubChem CID 69035
IUPAC Name propyl 4-methylbenzenesulfonate
SMILES CCCOS(=O)(=O)C1=CC=C(C=C1)C

Specifications

Color Yellow
Density 1.1500g/mL
Boiling Point 140.0°C (2.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4SO3(CH2)2CH3
Refractive Index 1.5065 to 1.5085
Quantity 25 g
Specific Gravity 1.15
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol
Formula Weight 214.28
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Propyl p-toluenesulfonate
Show More Show Less
compliance-icons
Product Identifier
  • Propyl p-toluenesulfonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.