Learn More
PRPF38A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24946-1-AP-150UL
 View more versions of this product
Product Code. 16893465
  • 410.00€ / 150µL
Description
PRPF38A is a protein coding gene involved in pre-mRNA splicing as a component of the spliceosome.
Specifications
Specifications
PRPF38A
Polyclonal
Unconjugated
PRPF38A
Pre mRNA splicing factor 38A, PRPF38A, RP5 965L7.1
Rabbit
Antigen Affinity Chromatography
RUO
84950
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8NAV1
PRPF38A
PRPF38A Fusion Protein Ag20980
150 μL