missing translation for 'onlineSavingsMsg'
Learn More

PRPF38A Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Specifications

Antigen PRPF38A
Concentration 0.2 mg/mL
Applications Immunofluorescence, Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16803475
View Documents
Proteintech
24946-1-AP-20UL
20 μL
145.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16893465
View Documents
Proteintech
24946-1-AP-150UL
150 μL
410.00€
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PRPF38A is a protein coding gene involved in pre-mRNA splicing as a component of the spliceosome.
Specifications

Specifications

PRPF38A
Immunofluorescence, Immunocytochemistry, Western Blot
Unconjugated
Rabbit
Human
Q8NAV1
84950
PRPF38A Fusion Protein Ag20980
Primary
-20°C
PRPF38A
0.2 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Pre mRNA splicing factor 38A, PRPF38A, RP5 965L7.1
PRPF38A
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.