missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PRRX1 Polyclonal specifically detects PRRX1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
Specifications
| Classification | Polyclonal |
| Quantity | 0.1 mg |
| Regulatory Status | RUO |
| Content And Storage | Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?