Learn More
PSMB5 Mouse anti-Human, Unconjugated, Clone: 2E9H1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67959-1-IG-20UL
Description
The PSMB5 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human PSMB5. This antibody recognizes human antigen. The PSMB5 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| PSMB5 | |
| Monoclonal | |
| Unconjugated | |
| 20 μL | |
| 5693 | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| 2E9H1 | |
| Mouse | |
| Primary | |
| Human |