Learn More
PSMB5 Mouse anti-Human, Unconjugated, Clone: 2E9H1, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | PSMB5 |
|---|---|
| Clone | 2E9H1 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17581466
|
Proteintech
67959-1-IG-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17571466
|
Proteintech
67959-1-IG-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PSMB5 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human PSMB5. This antibody recognizes human antigen. The PSMB5 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| PSMB5 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Human | |
| IgG2a |
| 2E9H1 | |
| Monoclonal | |
| Mouse | |
| 5693 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.