missing translation for 'onlineSavingsMsg'
Learn More

PST1 Antibody, HRP, Novus Biologicals™

Product Code. 18602016 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18602016 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18602016 Supplier Novus Biologicals Supplier No. NBP241063H
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PST1 Polyclonal antibody specifically detects PST1 in Yeast samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PST1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias Cell wall mannoprotein PST1, D4214, Haze protective factor 2, HPF2, YD9609.09, YDR055W
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide from the amino terminus of yeast Pst1. The immunogen is located within amino acids 80 - 130 of Pst1. Amino Acid Squence: DSLESITDSLNLQSLT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 851625
Target Species Yeast
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.